Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
ftr61
Entrez Gene ID
570050
Gene Name
finTRIM family, member 61
Gene Aliases
zgc:162037
Chromosome Location
Chr.24: 6034596 - 6038873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC116593.1 - - 1178 70
BC134866.1 - - 1251 70

View Full Product Details