Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tcta
Entrez Gene ID
436858
Gene Name
T cell leukemia translocation altered
Gene Aliases
wu:fb06g04, zgc:92721
Chromosome Location
Chr.6: 43015916 - 43019222 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002585.2 NP_001002585.1 1 - 2 428 61
GenBank mRNA AI397200.1 - 1 - 2 312 61
BC076201.1 - 1 - 2 336 61
CT633277.2 - 2 - 3 638 61
EH556465.1 - 1 - 2 399 61

View Full Product Details