Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ppp1cb
Entrez Gene ID
100003223
Gene Name
protein phosphatase 1, catalytic subunit, beta isozyme
Gene Aliases
ik:tdsubc_2f2, si:zc214p16.4, wu:fa09h01, wu:fa11e06, xx:tdsubc_2f2
Chromosome Location
Chr.17: 25635953 - 25649077 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004527.3 NP_001004527.2 2 - 3 205 93
GenBank mRNA BC095825.1 - 2 - 3 203 93
BC116539.1 - 2 - 3 205 93

View Full Product Details