Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl2
Entrez Gene ID
449774
Gene Name
ADP-ribosylation factor-like 2
Gene Aliases
zgc:103658
Chromosome Location
Chr.7: 20959 - 23100 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005947.1 NP_001005947.1 4 - 5 539 87
GenBank mRNA BC083457.1 - 4 - 5 539 87
BC153432.1 - 4 - 5 539 87

View Full Product Details