Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ing5b
Entrez Gene ID
368450
Gene Name
inhibitor of growth family, member 5b
Gene Aliases
ING1, p33, si:busm1-40g1.1, si:dz40g1.1, wu:fc94a02, zgc:100910
Chromosome Location
Chr.2: 36605994 - 36608760 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007168.1 NP_001007169.1 5 - 6 631 72
XM_005171273.4 XP_005171330.1 5 - 6 635 72
GenBank mRNA BC079501.1 - 5 - 6 631 72

View Full Product Details