Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
neil3
Entrez Gene ID
492463
Gene Name
nei-like DNA glycosylase 3
Gene Aliases
zgc:103553
Chromosome Location
Chr.14: 186584 - 204932 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007335.1 NP_001007336.1 1 - 2 212 128
XM_005157318.3 XP_005157375.1 1 - 2 271 128
GenBank mRNA BC085550.1 - 1 - 2 212 128
BC155157.1 - 1 - 2 192 128
BC164271.1 - 1 - 2 144 128

View Full Product Details