Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
neil3
Entrez Gene ID
492463
Gene Name
nei-like DNA glycosylase 3
Gene Aliases
zgc:103553
Chromosome Location
Chr.14: 186584 - 204932 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007335.1 NP_001007336.1 9 - 10 1570 75
XM_005157318.3 XP_005157375.1 9 - 10 1683 75
GenBank mRNA BC085550.1 - 9 - 10 1570 75
BC155157.1 - 9 - 10 1604 75
BC164271.1 - 9 - 10 1502 75

View Full Product Details