Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mtg1
Entrez Gene ID
100150980
Gene Name
mitochondrial ribosome-associated GTPase 1
Gene Aliases
gtpbpl, zgc:109944, zgc:86745
Chromosome Location
Chr.13: 21674445 - 21682873 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002140.1 NP_001002140.1 6 - 7 578 77
NM_001144056.1 NP_001137528.1 3 - 4 208 77
XM_009307279.3 XP_009305554.1 6 - 7 548 77
XM_021480487.1 XP_021336162.1 6 - 7 536 77
GenBank mRNA BC071417.1 - 6 - 7 578 77
BC095071.1 - 5 - 6 621 77
BC165080.1 - 3 - 4 205 77
BC171457.1 - 3 - 4 208 77

View Full Product Details