Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnajc9
Entrez Gene ID
436706
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 9
Gene Aliases
zgc:92648
Chromosome Location
Chr.12: 233882 - 243917 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002433.1 NP_001002433.1 5 - 6 813 74
GenBank mRNA BC076133.1 - 5 - 6 813 74

View Full Product Details