Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gatd1
Entrez Gene ID
100148360
Gene Name
glutamine amidotransferase like class 1 domain containing 1
Gene Aliases
pddc1, zgc:123196
Chromosome Location
Chr.25: 3801227 - 3808635 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037118.1 NP_001032195.1 3 - 4 432 83
GenBank mRNA BC107609.1 - 3 - 4 432 83

View Full Product Details