Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
foxred2
Entrez Gene ID
767722
Gene Name
FAD-dependent oxidoreductase domain containing 2
Gene Aliases
zgc:153216
Chromosome Location
Chr.3: 26019387 - 26032715 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076660.1 NP_001070128.1 4 - 5 906 77
XM_005163913.4 XP_005163970.1 4 - 5 935 77
XM_009299432.3 XP_009297707.1 4 - 5 1049 77
GenBank mRNA BC124275.1 - 4 - 5 906 77

View Full Product Details