Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
phkg1a
Entrez Gene ID
565379
Gene Name
phosphorylase kinase, gamma 1a (muscle)
Gene Aliases
zgc:153009
Chromosome Location
Chr.15: 25682 - 45683 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077612.1 NP_001071080.1 4 - 5 479 154
XM_021481442.1 XP_021337117.1 4 - 5 507 154
GenBank mRNA BC125847.1 - 4 - 5 479 154
BC164489.1 - 3 - 4 347 154

View Full Product Details