Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rhcga
Entrez Gene ID
100038797
Gene Name
Rh family, C glycoprotein a
Gene Aliases
rhcg2b, zgc:162132
Chromosome Location
Chr.7: 14291349 - 14302716 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001089577.1 NP_001083046.1 5 - 6 912 126
GenBank mRNA AB286867.1 - 5 - 6 937 126
BC139524.1 - 5 - 6 912 126

View Full Product Details