Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
valopb
Entrez Gene ID
560921
Gene Name
vertebrate ancient long opsin b
Gene Aliases
bZ1C10.4, si:rp71-1c10.4, va2, valop
Chromosome Location
Chr.12: 80935 - 98462 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110280.1 NP_001103750.1 2 - 3 644 106
GenBank mRNA AY996588.1 - 2 - 3 644 106
KT008403.1 - 2 - 3 562 106

View Full Product Details