Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
il13ra1
Entrez Gene ID
799475
Gene Name
interleukin 13 receptor, alpha 1
Gene Aliases
il-13ra1, si:ch211-150g2.2
Chromosome Location
Chr.7: 19153995 - 19168324 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110837.1 NP_001104307.1 4 - 5 642 101
GenBank mRNA BC162934.1 - 4 - 5 645 101

View Full Product Details