Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ca5a
Entrez Gene ID
569989
Gene Name
carbonic anhydrase Va
Gene Aliases
ca5, zgc:171653
Chromosome Location
Chr.25: 12794980 - 12809363 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111201.1 NP_001104671.1 4 - 5 963 76
XM_005163076.4 XP_005163133.1 4 - 5 76
GenBank mRNA BC153994.1 - 4 - 5 963 76

View Full Product Details