Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
map7d1b
Entrez Gene ID
796270
Gene Name
MAP7 domain containing 1b
Gene Aliases
zgc:172238
Chromosome Location
Chr.19: 3929084 - 4010263 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113635.1 NP_001107107.1 13 - 14 1851 100
XM_005173746.1 XP_005173803.1 15 - 16 2240 100
XM_005173747.1 XP_005173804.1 14 - 15 2224 100
XM_005173748.1 XP_005173805.1 14 - 15 2135 100
XM_009293978.3 XP_009292253.1 15 - 16 2240 100
GenBank mRNA BC154607.1 - 13 - 14 1851 100

Gene Symbol
LOC566136
Entrez Gene ID
566136
Gene Name
MAP7 domain-containing protein 1-like
Gene Aliases
-
Chromosome Location
Chr.14: 11307214 - 11335153 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_021481125.1 XP_021336800.1 1 - 2 55 100

Gene Symbol
LOC796866
Entrez Gene ID
796866
Gene Name
MAP7 domain-containing protein 1-like
Gene Aliases
-
Chromosome Location
Chr.14: 11388813 - 11392422 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_009307776.2 XP_009306051.1 2 - 3 200 100
XM_009307778.2 XP_009306053.1 1 - 2 55 100
XM_009307779.3 XP_009306054.1 2 - 3 200 100
XM_009307780.3 XP_009306055.1 1 - 2 56 100

Gene Symbol
si:ch73-281i18.3
Entrez Gene ID
100330519
Gene Name
si:ch73-281i18.3
Gene Aliases
-
Chromosome Location
Chr.19: 3890109 - 3896575 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002661011.5 XP_002661057.1 3 - 4 500 100
XM_009293983.1 XP_009292258.1 1 - 2 55 100

Gene Symbol
si:ch73-281i18.4
Entrez Gene ID
100318116
Gene Name
si:ch73-281i18.4
Gene Aliases
-
Chromosome Location
Chr.19: 3899887 - 3906372 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001366053.1 NP_001352982.1 3 - 4 510 100
XM_009293982.1 XP_009292257.1 1 - 2 55 100

Gene Symbol
si:ch73-281i18.6
Entrez Gene ID
100330842
Gene Name
si:ch73-281i18.6
Gene Aliases
-
Chromosome Location
Chr.19: 3919379 - 3925794 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002661016.5 XP_002661062.1 3 - 4 417 100

Gene Symbol
si:ch73-281i18.7
Entrez Gene ID
100535123
Gene Name
si:ch73-281i18.7
Gene Aliases
-
Chromosome Location
Chr.19: 3909622 - 3916076 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003200526.4 XP_003200574.1 3 - 4 447 100
XM_009293981.1 XP_009292256.1 1 - 2 55 100

View Full Product Details