Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
map7d1b
Entrez Gene ID
796270
Gene Name
MAP7 domain containing 1b
Gene Aliases
zgc:172238
Chromosome Location
Chr.19: 3929084 - 4010263 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113635.1 NP_001107107.1 14 - 15 1903 125
XM_005173746.1 XP_005173803.1 16 - 17 2292 125
XM_005173747.1 XP_005173804.1 15 - 16 2276 125
XM_005173748.1 XP_005173805.1 15 - 16 2187 125
GenBank mRNA BC154607.1 - 14 - 15 1903 125

View Full Product Details