Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ncalda
Entrez Gene ID
556178
Gene Name
neurocalcin delta a
Gene Aliases
zgc:175182
Chromosome Location
Chr.16: 9833246 - 9897580 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114410.1 NP_001107882.1 3 - 4 552 83
XM_005173483.4 XP_005173540.1 2 - 3 613 83
GenBank mRNA BC155337.1 - 3 - 4 552 83

View Full Product Details