Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cpne1
Entrez Gene ID
324841
Gene Name
copine I
Gene Aliases
cpne3, cpne3l, wu:fb69d07, wu:fc45a04, zgc:55873
Chromosome Location
Chr.11: 26375972 - 26401677 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199699.2 NP_955993.2 8 - 9 862 62
XM_005169258.4 XP_005169315.1 8 - 9 817 62
XM_005169262.3 XP_005169319.1 8 - 9 851 62
GenBank mRNA AY423010.1 - 8 - 9 797 62
BC050240.1 - 8 - 9 855 62
BC164658.1 - 7 - 8 765 62

View Full Product Details