Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dnajc17
Entrez Gene ID
393216
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 17
Gene Aliases
zgc:56091
Chromosome Location
Chr.17: 1042963 - 1174960 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200246.2 NP_956540.2 1 - 2 203 68
GenBank mRNA BC047828.1 - 1 - 2 107 68
BC065618.1 - 1 - 2 144 68

View Full Product Details