Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
stox1
Entrez Gene ID
568146
Gene Name
storkhead box 1
Gene Aliases
sb:cb624, zgc:174575
Chromosome Location
Chr.13: 18471546 - 18503983 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001123010.1 NP_001116482.1 1 - 2 477 110
GenBank mRNA BC134047.1 - 1 - 2 110
BC161651.1 - 1 - 2 477 110
BC167966.1 - 1 - 2 110

View Full Product Details