Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kcmf1
Entrez Gene ID
393927
Gene Name
potassium channel modulatory factor 1
Gene Aliases
zgc:64160
Chromosome Location
Chr.5: 55719757 - 55752201 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_200952.2 NP_957246.1 5 - 6 755 59
GenBank mRNA AY394941.1 - 5 - 6 755 59
BC053288.1 - 5 - 6 721 59

View Full Product Details