Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nek2
Entrez Gene ID
394025
Gene Name
NIMA-related kinase 2
Gene Aliases
zgc:55602
Chromosome Location
Chr.20: 13883165 - 13893910 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201050.1 NP_957344.1 2 - 3 237 102
GenBank mRNA BC048055.1 - 2 - 3 237 102
BC164531.1 - 1 - 2 135 102

View Full Product Details