Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ndel1b
Entrez Gene ID
333957
Gene Name
nudE neurodevelopment protein 1-like 1b
Gene Aliases
wu:fi23c04, wu:fi23e06
Chromosome Location
Chr.3: 27956 - 56619 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_201307.2 NP_958464.1 5 - 6 613 66
GenBank mRNA BC044386.2 - - 612 66
BC165239.1 - 4 - 5 560 66
EF133515.1 - 5 - 6 556 66

View Full Product Details