Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ttn.2
- Entrez Gene ID
-
317731
- Gene Name
- titin, tandem duplicate 2
- Gene Aliases
-
si:busm1-258d18.1, si:dz258d18.1, ttn, ttna, wu:fi04b11, wu:fi20g08
- Chromosome Location
-
Chr.9: 42895816 - 43073959 on Build GRCz11
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
CT652524.1 |
- |
3 - 4 |
781 |
87 |
- Gene Symbol
- ttn.1
- Entrez Gene ID
-
100001684
- Gene Name
- titin, tandem duplicate 1
- Gene Aliases
-
TTN, hm:zeh1256r, im:6911926, im:7146708, si:dZ167C3.1, si:dz249n21.1, ttnb, ttnl, wu:fc09a12
- Chromosome Location
-
Chr.9: 42732992 - 42873861 on Build GRCz11
View Full Product Details