Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
chmp2a
Entrez Gene ID
798901
Gene Name
charged multivesicular body protein 2A
Gene Aliases
bc2, fj10d05, wu:fi18h03, wu:fj10d05, zgc:55361
Chromosome Location
Chr.12: 27027314 - 27031031 on Build GRCz11
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_213517.1 NP_998682.1 4 - 5 619 70
GenBank mRNA BC045320.1 - 4 - 5 619 70
BC152230.1 - 4 - 5 619 70
BC171414.1 - 4 - 5 529 70

View Full Product Details