The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_214688.1 | NP_999853.1 | 8 - 9 | 3826 | 64 |
GenBank mRNA | AB119257.1 | - | 8 - 9 | 3826 | 64 |
BC162605.1 | - | 8 - 9 | 3812 | 64 |