Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NOS2
Entrez Gene ID
791246
Gene Name
nitric oxide synthase 2
Gene Aliases
INOS
Chromosome Location
Chr.11: 42217303 - 42253897 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081769.1 NP_001075238.1 5 - 6 626 76
XM_023651656.1 XP_023507424.1 - 626 76
XM_023651657.1 XP_023507425.1 - 1012 76
GenBank mRNA AY027883.1 - 5 - 6 626 76

View Full Product Details