Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SULT1E1
Entrez Gene ID
100034106
Gene Name
sulfotransferase family 1E member 1
Gene Aliases
-
Chromosome Location
Chr.3: 66828237 - 66851861 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081918.1 NP_001075387.1 6 - 7 800 111
XM_014738509.2 XP_014593995.2 - 919 111
GenBank mRNA DQ418452.1 - 6 - 7 800 111

View Full Product Details