Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNLY
Entrez Gene ID
100034133
Gene Name
granulysin
Gene Aliases
NKL
Chromosome Location
Chr.15: 19476760 - 19481087 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081929.3 NP_001075398.2 1 - 2 84 74
GenBank mRNA AF538056.3 - 1 - 2 84 74

View Full Product Details