Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNFSF13
Entrez Gene ID
100147233
Gene Name
TNF superfamily member 13
Gene Aliases
TNLG7B
Chromosome Location
Chr.11: 50873496 - 50877180 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205161.1 NP_001192090.1 1 - 2 999 84
XM_014736161.2 XP_014591647.1 - 1289 84
GenBank mRNA GU982935.1 - 1 - 2 262 84

View Full Product Details