Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBA52
Entrez Gene ID
100071005
Gene Name
ubiquitin A-52 residue ribosomal protein fusion product 1
Gene Aliases
-
Chromosome Location
Chr.21: 3829145 - 3831844 on Build EquCab3.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001242453.1 NP_001229382.1 4 - 5 334 95
XM_005604084.3 XP_005604141.1 - 438 95
XM_005604085.3 XP_005604142.1 - 367 95
XM_014734740.2 XP_014590226.1 - 393 95
GenBank mRNA CD464238.1 - 4 - 5 333 95
CD467165.1 - 3 - 4 278 95
CD536556.1 - 4 - 5 335 95

View Full Product Details