Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPS33
Entrez Gene ID
418119
Gene Name
mitochondrial ribosomal protein S33
Gene Aliases
-
Chromosome Location
Chr.1: 57013380 - 57019547 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015285566.2 XP_015141052.1 1 - 2 52 60
GenBank mRNA BX933786.1 - 1 - 2 24 60

View Full Product Details