Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OSGEPL1
Entrez Gene ID
423982
Gene Name
O-sialoglycoprotein endopeptidase like 1
Gene Aliases
-
Chromosome Location
Chr.7: 340872 - 352635 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004942397.3 XP_004942454.1 4 - 5 1177 89
XM_004942398.3 XP_004942455.1 4 - 5 1177 89
XM_004942399.3 XP_004942456.1 3 - 4 931 89
XM_025152269.1 XP_025008037.1 4 - 5 1177 89
GenBank mRNA BX930694.2 - 2 - 3 384 89
BX930963.2 - 2 - 3 1082 89
BX934991.1 - 4 - 5 961 89

View Full Product Details