Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MGME1
Entrez Gene ID
416719
Gene Name
mitochondrial genome maintenance exonuclease 1
Gene Aliases
C20orf72, C3H20ORF72, DDK1, MTDPS11, bA504H3.4
Chromosome Location
Chr.3: 15560641 - 15568616 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277518.1 NP_001264447.1 1 - 2 274 117
GenBank mRNA BU355182.1 - 1 - 2 274 117
BX929403.2 - 1 - 2 259 117
BX932303.1 - 1 - 2 274 117
BX933139.1 - 1 - 2 239 117

View Full Product Details