Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GAL
Entrez Gene ID
423117
Gene Name
galanin and GMAP prepropeptide
Gene Aliases
-
Chromosome Location
Chr.5: 16762606 - 16770584 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145389.2 NP_001138861.1 5 - 6 467 88
NM_001159678.1 NP_001153150.1 6 - 7 539 88
GenBank mRNA BU265315.1 - 5 - 6 467 88
BX930206.2 - 5 - 6 437 88
CR386974.1 - 5 - 6 467 88
FJ619535.1 - 5 - 6 328 88
FJ848872.1 - 6 - 7 400 88

View Full Product Details