Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2H
Entrez Gene ID
424954
Gene Name
RNA polymerase II subunit H
Gene Aliases
-
Chromosome Location
Chr.9: 16103738 - 16105679 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277465.1 NP_001264394.1 3 - 4 289 68
NM_001277467.1 NP_001264396.1 2 - 3 205 68
GenBank mRNA BX929949.2 - 2 - 3 200 68
BX931358.2 - 3 - 4 281 68
CF256811.1 - 3 - 4 291 68
CR386803.1 - 3 - 4 267 68

View Full Product Details