Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLR2H
Entrez Gene ID
424954
Gene Name
RNA polymerase II subunit H
Gene Aliases
-
Chromosome Location
Chr.9: 16103738 - 16105679 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277465.1 NP_001264394.1 4 - 5 368 217
NM_001277467.1 NP_001264396.1 3 - 4 284 217
GenBank mRNA BX929949.2 - 3 - 4 279 217
BX931358.2 - 4 - 5 360 217
BX933533.2 - 4 - 5 250 133
CF256811.1 - 4 - 5 370 217
CR386803.1 - 4 - 5 346 217

View Full Product Details