Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN2
Entrez Gene ID
422292
Gene Name
claudin 2
Gene Aliases
-
Chromosome Location
Chr.4: 10939321 - 10941847 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277622.1 NP_001264551.1 1 - 2 183 125
GenBank mRNA BX934932.2 - 1 - 2 183 125

View Full Product Details