Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB40B
Entrez Gene ID
417338
Gene Name
RAB40B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.18: 3365331 - 3388901 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278050.3 NP_001264979.1 2 - 3 544 71
GenBank mRNA BX950715.2 - 2 - 3 544 71

View Full Product Details