Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSTF1
Entrez Gene ID
419331
Gene Name
cleavage stimulation factor subunit 1
Gene Aliases
-
Chromosome Location
Chr.20: 12011422 - 12018869 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_025142431.1 XP_024998199.1 2 - 3 364 88
GenBank mRNA BX929902.2 - 2 - 3 381 88
CR385371.1 - 3 - 4 540 88

View Full Product Details