Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL26A1
Entrez Gene ID
417504
Gene Name
collagen type XXVI alpha 1 chain
Gene Aliases
EMID2
Chromosome Location
Chr.19: 3659652 - 3832077 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004946472.3 XP_004946529.1 1 - 2 320 120
XM_015295986.2 XP_015151472.1 1 - 2 321 120
XM_015295987.2 XP_015151473.1 1 - 2 321 120
XM_015295988.2 XP_015151474.1 1 - 2 321 120
XM_015295989.2 XP_015151475.1 1 - 2 320 120
XM_015295990.2 XP_015151476.1 1 - 2 320 120
GenBank mRNA CR387598.1 - 1 - 2 97 120

View Full Product Details