Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAXBP1
Entrez Gene ID
418499
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
Chromosome Location
Chr.1: 106415249 - 106452091 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_416707.6 XP_416707.3 16 - 17 7403 93
GenBank mRNA CR386728.1 - 2 - 3 111 93

View Full Product Details