Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLDN15
Entrez Gene ID
424768
Gene Name
claudin 15
Gene Aliases
claudin-15, claudin-19
Chromosome Location
Chr.9: 2841999 - 2861738 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR406782.1 - 4 - 5 569 67

View Full Product Details