Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KMT2C
Entrez Gene ID
420437
Gene Name
lysine methyltransferase 2C
Gene Aliases
MLL3
Chromosome Location
Chr.2: 6620024 - 6808024 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004939143.3 XP_004939200.1 48 - 49 13132 102
XM_015281218.2 XP_015136704.1 49 - 50 13026 102
XM_015281219.2 XP_015136705.1 48 - 49 13129 102
XM_015281220.2 XP_015136706.1 48 - 49 13129 102
XM_015281221.2 XP_015136707.1 48 - 49 13111 102
XM_015281222.2 XP_015136708.1 47 - 48 13083 102
XM_015281223.2 XP_015136709.1 47 - 48 13065 102
XM_015281224.2 XP_015136710.1 48 - 49 13059 102
XM_015281225.2 XP_015136711.1 47 - 48 13011 102
XM_015281226.2 XP_015136712.1 47 - 48 12967 102
XM_015281227.2 XP_015136713.1 48 - 49 12463 102
XM_015281228.2 XP_015136714.1 46 - 47 12852 102
XM_015281229.2 XP_015136715.1 46 - 47 12756 102
XM_015281230.2 XP_015136716.1 46 - 47 12919 102
XM_025148296.1 XP_025004064.1 48 - 49 13108 102
XM_025148298.1 XP_025004066.1 48 - 49 12251 102
XM_025148299.1 XP_025004067.1 49 - 50 12571 102
GenBank mRNA CR733145.1 - 1 - 2 185 102

View Full Product Details