Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTGS2
Entrez Gene ID
396451
Gene Name
prostaglandin-endoperoxide synthase 2
Gene Aliases
CEF147, PGHS2, PHSII
Chromosome Location
Chr.8: 10714136 - 10722126 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167718.1 NP_001161190.1 2 - 3 675 108
NM_001167719.1 NP_001161191.1 2 - 3 246 108
GenBank mRNA CF250042.1 - 2 - 3 246 108
M64990.1 - 2 - 3 771 108

View Full Product Details