Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKR7A2
Entrez Gene ID
419471
Gene Name
aldo-keto reductase family 7, member A2 (aflatoxin aldehyde reductase)
Gene Aliases
-
Chromosome Location
Chr.21: 4676380 - 4680371 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_417628.6 XP_417628.2 1 - 2 210 134
GenBank mRNA BX930804.2 - 1 - 2 83 134
BX930844.1 - 1 - 2 200 134
BX931408.1 - 1 - 2 179 134
BX934026.2 - 1 - 2 202 134
BX934046.1 - 1 - 2 202 134
CR353499.1 - 1 - 2 197 134

View Full Product Details