Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRKX
Entrez Gene ID
418656
Gene Name
protein kinase, X-linked
Gene Aliases
-
Chromosome Location
Chr.1: 129430051 - 129485175 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015277728.2 XP_015133214.1 7 - 8 1289 150
GenBank mRNA CR354088.1 - 7 - 8 886 150

View Full Product Details