Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PTTG2
Entrez Gene ID
416158
Gene Name
pituitary tumor-transforming 2
Gene Aliases
PTTG1
Chromosome Location
Chr.13: 8539665 - 8542809 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277740.1 NP_001264669.1 3 - 4 505 97
GenBank mRNA AJ441614.1 - 3 - 4 338 97
BX932212.2 - 3 - 4 404 97
CN232875.1 - 3 - 4 506 97

View Full Product Details